Assay Details
| Gene Symbol: CHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966985.2 | XP_013822439.1 | 2 - 3 | 426 | 55 | |
| GenBank mRNA | KA348515.1 | - | 2 - 3 | 409 | 55 | |
More Information
Set Membership: |
|

CHAC1
Genomic Map