Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967703.2 | XP_013823157.2 | 7 - 8 | 1700 | 87 | |
| XM_013967707.2 | XP_013823161.2 | 7 - 8 | 1666 | 87 | ||
| GenBank mRNA | JO426720.1 | - | 1 - 2 | 1333 | 87 | |
More Information
Set Membership: |
|

NAB1
Genomic Map