Assay Details
| Gene Symbol: TAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057598.1 | XP_017913087.1 | 2 - 3 | 1500 | 61 | |
| GenBank mRNA | KA341020.1 | - | 2 - 3 | 321 | 61 | |
More Information
Set Membership: |
|

TAF4
Genomic Map