Assay Details
| Gene Symbol: LOC106503078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061697.1 | XP_017917186.1 | 15 - 16 | 2335 | 57 | |
| XM_018061698.1 | XP_017917187.1 | 15 - 16 | 2162 | 57 | ||
| XM_018061699.1 | XP_017917188.1 | 14 - 15 | 2031 | 57 | ||
| GenBank mRNA | JO349596.2 | - | 4 - 5 | 973 | 57 | |
More Information
Set Membership: |
|

LOC106503078
Genomic Map