Assay Details
| Gene Symbol: CMTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062087.1 | XP_017917576.1 | 3 - 4 | 731 | 57 | |
| GenBank mRNA | JO426150.1 | - | 3 - 4 | 735 | 57 | |
More Information
Set Membership: |
|

CMTM4
Genomic Map