Assay Details
| Gene Symbol: RFLNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064375.1 | XP_017919864.1 | 2 - 3 | 402 | 62 | |
| GenBank mRNA | JO585569.2 | - | 2 - 3 | 417 | 62 | |
More Information
Set Membership: |
|

RFLNB
Genomic Map