Assay Details
| Gene Symbol: OGFOD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065401.1 | XP_017920890.1 | 2 - 3 | 404 | 59 | |
| GenBank mRNA | JO587059.2 | - | 10 - 11 | 1781 | 59 | |
More Information
Set Membership: |
|

OGFOD3
Genomic Map