Assay Details
| Gene Symbol: LOC102172305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066738.1 | XP_017922227.1 | 13 - 14 | 1684 | 93 | |
| GenBank mRNA | JO343663.2 | - | 6 - 7 | 666 | 93 | |
More Information
Set Membership: |
|

LOC102172305
Genomic Map