Assay Details
| Gene Symbol: LOC108638604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066739.1 | XP_017922228.1 | 225 | 80 | ||
| GenBank mRNA | JO498516.2 | - | 1614 | 80 | ||
| Gene Symbol: LOC102172305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066738.1 | XP_017922227.1 | 4402 | 80 | ||
| Gene Symbol: LOC108638594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066675.1 | XP_017922164.1 | 333 | 80 | ||
More Information
Set Membership: |
|

LOC108638604
Genomic Map