Assay Details
| Gene Symbol: DDAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038620.1 | XP_017894109.1 | 4 - 5 | 774 | 77 | |
| XM_018038621.1 | XP_017894110.1 | 4 - 5 | 1213 | 77 | ||
| XM_018038622.1 | XP_017894111.1 | 4 - 5 | 1179 | 77 | ||
| GenBank mRNA | JR126569.1 | - | 3439 | 77 | ||
More Information
Set Membership: |
|

DDAH2
Genomic Map