Assay Details
| Gene Symbol: NME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040332.1 | XP_017895821.1 | 3 - 4 | 434 | 79 | |
| XM_018040333.1 | XP_017895822.1 | 3 - 4 | 567 | 79 | ||
| XM_018040334.1 | XP_017895823.1 | 3 - 4 | 461 | 79 | ||
| GenBank mRNA | JO577832.1 | - | 3 - 4 | 459 | 79 | |
| JT685917.1 | - | 4 - 5 | 484 | 79 | ||
More Information
Set Membership: |
|

NME4
Genomic Map