Assay Details
| Gene Symbol: ZNF316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040816.1 | XP_017896305.1 | 2 - 3 | 421 | 48 | |
| GenBank mRNA | JO435424.2 | - | 4 - 5 | 967 | 48 | |
More Information
Set Membership: |
|

ZNF316
Genomic Map