Assay Details
| Gene Symbol: LOC102181832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040400.1 | XP_017895889.1 | 8 - 9 | 1155 | 60 | |
| GenBank mRNA | JO360672.2 | - | 8 - 9 | 1524 | 60 | |
| JO504852.2 | - | 2 - 3 | 131 | 60 | ||
More Information
Set Membership: |
|

LOC102181832
Genomic Map