Assay Details
| Gene Symbol: LOC102181832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040400.1 | XP_017895889.1 | 9 - 10 | 1269 | 71 | |
| GenBank mRNA | JO360672.2 | - | 7 - 8 | 1410 | 71 | |
More Information
Set Membership: |
|

LOC102181832
Genomic Map