Assay Details
| Gene Symbol: PCYT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675060.3 | XP_005675117.1 | 6 - 7 | 718 | 72 | |
| XM_013963486.2 | XP_013818940.1 | 6 - 7 | 668 | 72 | ||
| GenBank mRNA | JO343300.2 | - | 2 - 3 | 294 | 72 | |
| JO345971.2 | - | 3 - 4 | 826 | 72 | ||
| JO372074.2 | - | 3 - 4 | 492 | 72 | ||
| JO398362.2 | - | 6 - 7 | 677 | 72 | ||
| JO435071.2 | - | 6 - 7 | 647 | 72 | ||
| JO445552.2 | - | 6 - 7 | 647 | 72 | ||
| JO514008.2 | - | 2 - 3 | 334 | 72 | ||
| JO541803.2 | - | 6 - 7 | 675 | 72 | ||
| JO541804.2 | - | 6 - 7 | 675 | 72 | ||
| JO541805.2 | - | 6 - 7 | 675 | 72 | ||
| JO574460.2 | - | 4 - 5 | 421 | 72 | ||
| JO574739.2 | - | 4 - 5 | 421 | 72 | ||
| JO604404.2 | - | 3 - 4 | 826 | 72 | ||
| JO618945.2 | - | 6 - 7 | 690 | 72 | ||
| JO618946.2 | - | 6 - 7 | 690 | 72 | ||
| KA348630.1 | - | 6 - 7 | 694 | 72 | ||
More Information
Set Membership: |
|

PCYT1A
Genomic Map