Assay Details
| Gene Symbol: HJV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677729.3 | XP_005677786.2 | 3 - 4 | 931 | 79 | |
| XM_005677730.3 | XP_005677787.2 | 3 - 4 | 949 | 79 | ||
| XM_013962460.2 | XP_013817914.2 | 2 - 3 | 742 | 79 | ||
| GenBank mRNA | JO411586.2 | - | 1 - 2 | 1281 | 79 | |
More Information
Set Membership: |
|

HJV
Genomic Map