Assay Details
| Gene Symbol: LOC102169641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678020.3 | XP_005678077.1 | 4 - 5 | 958 | 101 | |
| GenBank mRNA | JO535150.2 | - | 3 - 4 | 447 | 101 | |
More Information
Set Membership: |
|

LOC102169641
Genomic Map