Assay Details
| Gene Symbol: GSTK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679551.3 | XP_005679608.1 | 7 - 8 | 716 | 52 | |
| GenBank mRNA | JO441907.2 | - | 7 - 8 | 695 | 52 | |
More Information
Set Membership: |
|

GSTK1
Genomic Map