Assay Details
| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680006.3 | XP_005680063.1 | 11 - 12 | 972 | 66 | |
| GenBank mRNA | JO613294.2 | - | 11 - 12 | 949 | 66 | |
More Information
Set Membership: |
|

PRKAG1
Genomic Map