Assay Details
| Gene Symbol: DTX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048013.1 | XP_017903502.1 | 4 - 5 | 1246 | 72 | |
| XM_018048014.1 | XP_017903503.1 | 4 - 5 | 1261 | 72 | ||
| XM_018048015.1 | XP_017903504.1 | 4 - 5 | 1221 | 72 | ||
| XM_018048016.1 | XP_017903505.1 | 4 - 5 | 1237 | 72 | ||
More Information
Set Membership: |
|

DTX3
Genomic Map