Assay Details
| Gene Symbol: SNCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681428.3 | XP_005681485.1 | 5 - 6 | 512 | 70 | |
| XM_005681429.3 | XP_005681486.1 | 5 - 6 | 805 | 70 | ||
| XM_005681430.3 | XP_005681487.1 | 5 - 6 | 714 | 70 | ||
| XM_013964535.2 | XP_013819989.1 | 5 - 6 | 593 | 70 | ||
| XM_013964536.2 | XP_013819990.1 | 5 - 6 | 487 | 70 | ||
| XM_013964537.2 | XP_013819991.1 | 5 - 6 | 519 | 70 | ||
| XM_013964538.2 | XP_013819992.1 | 5 - 6 | 570 | 70 | ||
| XM_013964539.2 | XP_013819993.1 | 5 - 6 | 545 | 70 | ||
| XM_018049310.1 | XP_017904799.1 | 5 - 6 | 434 | 70 | ||
| GenBank mRNA | JO631885.2 | - | 1 - 2 | 320 | 70 | |
| KA322407.1 | - | 5 - 6 | 466 | 70 | ||
More Information
Set Membership: |
|

SNCA
Genomic Map