Assay Details
| Gene Symbol: TNFSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684351.3 | XP_005684408.1 | 2 - 3 | 532 | 97 | |
| GenBank mRNA | JO323200.2 | - | 3 - 4 | 2323 | 97 | |
| JO323201.2 | - | 3 - 4 | 2323 | 97 | ||
More Information
Set Membership: |
|

TNFSF8
Genomic Map