Assay Details
| Gene Symbol: SLC35A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053161.1 | XP_017908650.1 | 3 - 4 | 558 | 66 | |
| XM_018053162.1 | XP_017908651.1 | 3 - 4 | 562 | 66 | ||
More Information
Set Membership: |
|

SLC35A1
Genomic Map