Assay Details
| Gene Symbol: CRNKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057255.1 | XP_017912744.1 | 3 - 4 | 404 | 67 | |
| GenBank mRNA | JO639200.2 | - | 1 - 2 | 149 | 67 | |
More Information
Set Membership: |
|

CRNKL1
Genomic Map