Assay Details
| Gene Symbol: BCAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688743.3 | XP_005688800.2 | 2 - 3 | 445 | 63 | |
| XM_005688744.3 | XP_005688801.2 | 2 - 3 | 441 | 63 | ||
| XM_013968981.2 | XP_013824435.2 | 2 - 3 | 445 | 63 | ||
| XM_013968982.2 | XP_013824436.2 | 2 - 3 | 445 | 63 | ||
| XM_013968983.2 | XP_013824437.2 | 2 - 3 | 446 | 63 | ||
| XM_018058051.1 | XP_017913540.1 | 2 - 3 | 447 | 63 | ||
| XM_018058052.1 | XP_017913541.1 | 2 - 3 | 441 | 63 | ||
More Information
Set Membership: |
|

BCAS1
Genomic Map