Assay Details
| Gene Symbol: LOC102180514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689470.3 | XP_005689527.2 | 2 - 3 | 581 | 62 | |
| GenBank mRNA | JP351434.2 | - | 1 - 2 | 1015 | 62 | |
More Information
Set Membership: |
|

LOC102180514
Genomic Map