Assay Details
| Gene Symbol: LOC102181880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689475.3 | XP_005689532.2 | 2 - 3 | 233 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102181880| Gene Symbol: LOC102181880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689475.3 | XP_005689532.2 | 2 - 3 | 233 | 107 | |
Set Membership: |
|