Assay Details
| Gene Symbol: TBCEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689558.3 | XP_005689615.1 | 6 - 7 | 1336 | 64 | |
| XM_018059642.1 | XP_017915131.1 | 6 - 7 | 1032 | 64 | ||
| XM_018059643.1 | XP_017915132.1 | 6 - 7 | 922 | 64 | ||
| GenBank mRNA | JO604346.2 | - | 2 - 3 | 1666 | 64 | |
More Information
Set Membership: |
|

TBCEL
Genomic Map