Assay Details
| Gene Symbol: ROPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694858.3 | XP_005694915.1 | 2 - 3 | 269 | 64 | |
| GenBank mRNA | JO444597.2 | - | 4 - 5 | 651 | 64 | |
More Information
Set Membership: |
|

ROPN1L
Genomic Map