Assay Details
| Gene Symbol: CMTM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973449.2 | XP_013828903.1 | 3 - 4 | 346 | 71 | |
| XM_018038315.1 | XP_017893804.1 | 3 - 4 | 520 | 71 | ||
More Information
Set Membership: |
|

CMTM8
Genomic Map