Assay Details
| Gene Symbol: LOC102169925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039758.1 | XP_017895247.1 | 2 - 3 | 544 | 65 | |
| XM_018039759.1 | XP_017895248.1 | 2 - 3 | 340 | 65 | ||
| GenBank mRNA | JO584338.1 | - | 2 - 3 | 290 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018039759.1 transcripts.
Set Membership: |
|

LOC102169925
Genomic Map