Assay Details
| Gene Symbol: GPRC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040564.1 | XP_017896053.1 | 3 - 4 | 1263 | 71 | |
| GenBank mRNA | JO434014.2 | - | 3 - 4 | 1266 | 71 | |
More Information
Set Membership: |
|

GPRC5B
Genomic Map