Assay Details
| Gene Symbol: LOC102176495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699329.3 | XP_005699386.1 | 7 - 8 | 679 | 77 | |
| GenBank mRNA | KA315603.1 | - | 7 - 8 | 732 | 77 | |
More Information
Set Membership: |
|

LOC102176495
Genomic Map