Assay Details
| Gene Symbol: TRIM25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709548.3 | XP_005709605.3 | 5 - 6 | 1258 | 62 | |
| GenBank mRNA | JO577496.1 | - | 4 - 5 | 1237 | 62 | |
More Information
Set Membership: |
|

TRIM25
Genomic Map