Assay Details
Gene Symbol: LOC102179319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962506.2 | XP_013817960.1 | 6 - 7 | 897 | 82 | |
GenBank mRNA | JO632563.2 | - | 1 - 2 | 481 | 82 | |
JT658395.1 | - | 1 - 2 | 212 | 82 | ||
JT738675.1 | - | 2 - 3 | 287 | 82 | ||
KA347851.1 | - | 1 - 2 | 211 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
