Assay Details
Gene Symbol: LOC102179319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962506.2 | XP_013817960.1 | 8 - 9 | 1179 | 89 | |
GenBank mRNA | JO382243.2 | - | 1 - 2 | 434 | 89 | |
JO543593.2 | - | 2 - 3 | 193 | 89 | ||
JO632563.2 | - | 3 - 4 | 763 | 89 | ||
KA346920.1 | - | 2 - 3 | 193 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
