Assay Details
| Gene Symbol: SYPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047233.1 | XP_017902722.1 | 2 - 3 | 246 | 77 | |
| XM_018047234.1 | XP_017902723.1 | 2 - 3 | 296 | 77 | ||
| GenBank mRNA | JO536121.2 | - | 2 - 3 | 304 | 77 | |
More Information
Set Membership: |
|

SYPL1
Genomic Map