Assay Details
| Gene Symbol: ADAMTS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965221.2 | XP_013820675.2 | 2 - 3 | 2564 | 59 | |
| GenBank mRNA | JO439855.2 | - | 1 - 2 | 450 | 59 | |
| JO532636.2 | - | 4 - 5 | 726 | 59 | ||
| JO601700.2 | - | 1 - 2 | 256 | 59 | ||
| KA323386.1 | - | 7 - 8 | 2067 | 59 | ||
More Information
Set Membership: |
|

ADAMTS5
Genomic Map