Assay Details
| Gene Symbol: LSM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050771.1 | XP_017906260.1 | 3 - 4 | 209 | 70 | |
| GenBank mRNA | JT659320.1 | - | 2 - 3 | 207 | 70 | |
More Information
Set Membership: |
|

LSM7
Genomic Map