Assay Details
| Gene Symbol: DOP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052710.1 | XP_017908199.1 | 9 - 10 | 1308 | 71 | |
| XM_018052719.1 | XP_017908208.1 | 9 - 10 | 1308 | 71 | ||
| XM_018052727.1 | XP_017908216.1 | 9 - 10 | 1308 | 71 | ||
| XM_018052737.1 | XP_017908226.1 | 9 - 10 | 1308 | 71 | ||
More Information
Set Membership: |
|

DOP1B
Genomic Map