Assay Details
| Gene Symbol: TP53INP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057741.1 | XP_017913230.1 | 2 - 3 | 415 | 59 | |
| XM_018057742.1 | XP_017913231.1 | 3 - 4 | 525 | 59 | ||
| GenBank mRNA | JO425440.1 | - | 2 - 3 | 3718 | 59 | |
| JO633667.2 | - | 2 - 3 | 3697 | 59 | ||
More Information
Set Membership: |
|

TP53INP2
Genomic Map