Assay Details
| Gene Symbol: PTPRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058998.1 | XP_017914487.1 | 15 - 16 | 2473 | 57 | |
| XM_018059002.1 | XP_017914491.1 | 15 - 16 | 2274 | 57 | ||
More Information
Set Membership: |
|

PTPRN
Genomic Map