Assay Details
| Gene Symbol: GPD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038391.1 | XP_017893880.1 | 2 - 3 | 340 | 59 | |
| GenBank mRNA | JO441367.2 | - | 2 - 3 | 366 | 59 | |
| JT673693.1 | - | 2 - 3 | 366 | 59 | ||
More Information
Set Membership: |
|

GPD1L
Genomic Map