Assay Details
| Gene Symbol: WDR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066747.1 | XP_017922236.1 | 4 - 5 | 2967 | 51 | |
| GenBank mRNA | JO644712.2 | - | 5 - 6 | 1331 | 51 | |
More Information
Set Membership: |
|

WDR6
Genomic Map