Assay Details
| Gene Symbol: PDZRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067086.1 | XP_017922575.1 | 7 - 8 | 1833 | 66 | |
| XM_018067087.1 | XP_017922576.1 | 5 - 6 | 854 | 66 | ||
More Information
Set Membership: |
|

PDZRN3
Genomic Map