Assay Details
| Gene Symbol: LOC102169654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044218.1 | XP_017899707.1 | 16 - 17 | 2822 | 79 | |
| XM_018044219.1 | XP_017899708.1 | 17 - 18 | 2627 | 79 | ||
| XM_018044220.1 | XP_017899709.1 | 15 - 16 | 2711 | 79 | ||
| XM_018044221.1 | XP_017899710.1 | 16 - 17 | 2690 | 79 | ||
More Information
Set Membership: |
|

LOC102169654
Genomic Map