Assay Details
| Gene Symbol: LOC102169654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044218.1 | XP_017899707.1 | 7 - 8 | 1242 | 106 | |
| XM_018044219.1 | XP_017899708.1 | 8 - 9 | 1047 | 106 | ||
| XM_018044220.1 | XP_017899709.1 | 7 - 8 | 1242 | 106 | ||
| XM_018044221.1 | XP_017899710.1 | 7 - 8 | 1242 | 106 | ||
More Information
Set Membership: |
|

LOC102169654
Genomic Map