Assay Details
Gene Symbol: ATP6V1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674936.3 | XP_005674993.1 | 3 - 4 | 338 | 59 | |
XM_005674937.3 | XP_005674994.1 | 3 - 4 | 366 | 59 | ||
GenBank mRNA | JO324867.2 | - | 3 - 4 | 295 | 59 | |
JO324869.2 | - | 3 - 4 | 295 | 59 | ||
JO353936.2 | - | 1 - 2 | 162 | 59 | ||
JO353937.2 | - | 1 - 2 | 162 | 59 | ||
JO414927.2 | - | 12 - 13 | 2127 | 59 | ||
JO440140.2 | - | 5 - 6 | 799 | 59 | ||
JO441135.2 | - | 5 - 6 | 799 | 59 | ||
JO519447.2 | - | 4 - 5 | 548 | 59 | ||
JO564043.2 | - | 1 - 2 | 154 | 59 | ||
JO599584.2 | - | 12 - 13 | 3908 | 59 | ||
JO599585.2 | - | 12 - 13 | 3893 | 59 | ||
JO634192.2 | - | 12 - 13 | 3929 | 59 | ||
JT658415.1 | - | 1 - 2 | 108 | 59 | ||
JT705695.1 | - | 3 - 4 | 292 | 59 | ||
JT738697.1 | - | 3 - 4 | 274 | 59 | ||
KA312673.1 | - | 3 - 4 | 317 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
