Assay Details
| Gene Symbol: BORCS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682087.3 | XP_005682144.2 | 2 - 3 | 252 | 90 | |
| XM_005682088.3 | XP_005682145.2 | 2 - 3 | 252 | 90 | ||
| XM_005682089.3 | XP_005682146.2 | 2 - 3 | 252 | 90 | ||
| XM_005682090.3 | XP_005682147.2 | 2 - 3 | 252 | 90 | ||
More Information
Set Membership: |
|

BORCS8
Genomic Map