Assay Details
| Gene Symbol: LOC102181586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682151.3 | XP_005682208.1 | 11 - 12 | 1482 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102181586| Gene Symbol: LOC102181586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682151.3 | XP_005682208.1 | 11 - 12 | 1482 | 77 | |
Set Membership: |
|